Anti-MCTS1 Polyclonal Antibody

Supplier: Abnova
PAB12560
10725-158EA 638.29 USD
10725-158
Anti-MCTS1 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against Mcts1.

Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: MCTS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR