Anti-B2M Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B2M-01]

Supplier: Abnova
MAB5021
10692-658EA 329 USD
10692-658
Anti-B2M Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B2M-01]
Antibodies
Mouse monoclonal antibody raised against B2M.

Recommended Dilutions: Flow Cytometry: 1:200

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: B2M-01
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR