Anti-CORO1A Polyclonal Antibody

Supplier: Abnova
PAB12563
10708-828EA 746.19 USD
10708-828
Anti-CORO1A Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant CORO1A.

Recommended Dilutions: Immunofluorescence: 1:500-1:1000; Western Blot: 1:20; Immunocytochemistry-1:5000

Type: Primary
Antigen: CORO1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR