Anti-HLA-Class I Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-147]

Supplier: Abnova
MAB5024
10720-960EA 346 USD
10720-960
Anti-HLA-Class I Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-147]
Antibodies
Mouse monoclonal antibody raised against native HLA-Class I.

Recommended Dilutions: Flow Cytometry: 5 ?g/ml

Type: Primary
Antigen: HLA-Class I
Clonality: PE (Phycoerythrin)
Clone: Monoclonal
Conjugation: MEM-147
Epitope:
Host: IgG1
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR