Anti-Sir2.1 Polyclonal Antibody

Supplier: Abnova
PAB12566
10737-168EA 530.32 USD
10737-168
Anti-Sir2.1 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant sir-2.1.

Recommended Dilutions: Western Blot: 1:3000

Type: Primary
Antigen: Sir2.1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Nematoda
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR