Anti-JNK1 Polyclonal Antibody

Supplier: Abnova
PAB12568
10721-560EA 530.32 USD
10721-560
Anti-JNK1 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant jnk-1.

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: JNK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Nematoda
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR