Anti-PPP1R13B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20346
10729-886EA 428.55 USD
10729-886
Anti-PPP1R13B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant PPP1R13B.

Recommended Dilutions: Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: PPP1R13B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR