Anti-ACIN1 Polyclonal Antibody

Supplier: Abnova
PAB12575
10687-428EA 638.29 USD
10687-428
Anti-ACIN1 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ACIN1.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: ACIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR