Anti-PDIA2 Polyclonal Antibody

Supplier: Abnova
PAB12587
10728-752EA 530.32 USD
10728-752
Anti-PDIA2 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length native PDIA2.

Recommended Dilutions: Immunocytochemistry: 1:5000-1:20000; Immunohistochemistry (Frozen sections): 1:100-1:1000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100-1:500; Immunoprecipitation: 1-3 ?g; Western Blot: 1:10000-1:20000

Type: Primary
Antigen: PDIA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Bovine, Human, Mouse, Primates, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR