Anti-PTGFRN Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20920
10730-554EA 428.55 USD
10730-554
Anti-PTGFRN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant PTGFRN.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: PTGFRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR