Anti-CACHD1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20925
10704-622EA 428.55 USD
10704-622
Anti-CACHD1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CACHD1.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500

Type: Primary
Antigen: CACHD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR