Anti-CRABP1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20926
10709-022EA 428.55 USD
10709-022
Anti-CRABP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CRABP1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: CRABP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR