Anti-HBsAg Mouse Monoclonal Antibody [clone: P3G9A8]

Supplier: Abnova
MAB3541
10627-688EA 345.64 USD
10627-688
Anti-HBsAg Mouse Monoclonal Antibody [clone: P3G9A8]
Antibodies
Mouse monoclonal antibody raised against hepatitis B virus surface antigen.

Recommended Dilutions: Immunohistochemistry: 1:10-1:25

Type: Primary
Antigen: Hepatitis B virus surface antigen
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: P3G9A8
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR