Anti-papilloma virus early Mouse Monoclonal Antibody [clone: C1P5]

Supplier: Abnova
MAB3544
10638-038EA 428.55 USD
10638-038
Anti-papilloma virus early Mouse Monoclonal Antibody [clone: C1P5]
Antibodies
Mouse monoclonal antibody raised against recombinant human papilloma virus Early Protein 6 (Type 16, 18).

Type: Primary
Antigen: papilloma virus early
Clonality: Monoclonal
Clone: C1P5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR