Anti-BTK Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12936
10692-928EA 428 USD
10692-928
Anti-BTK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of BTK.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: BTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR