Anti-Melanoma Mouse Monoclonal Antibody [clone: NKI/C3]

Supplier: Abnova
MAB3552
10628-264EA 638 USD
10628-264
Anti-Melanoma Mouse Monoclonal Antibody [clone: NKI/C3]
Antibodies
Mouse monoclonal antibody raised against Melanoma.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: Melanoma
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: NKI/C3
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR