Anti-RPS6KA1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12946
10731-956EA 428 USD
10731-956
Anti-RPS6KA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of RPS6KA1.

Recommended Dilutions: Western Blot: 5-10 ?g/ml

Type: Primary
Antigen: RPS6KA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR