Anti-NRCAM Mouse Monoclonal Antibody [clone: RNL-1]

Supplier: Abnova
MAB3561
10628-368EA 747 USD
10628-368
Anti-NRCAM Mouse Monoclonal Antibody [clone: RNL-1]
Antibodies
Mouse monoclonal antibody raised against native NRCAM.

Recommended Dilutions: Immunohistochemistry: 1:10-1:25

Type: Primary
Antigen: NRCAM
Clonality: Monoclonal
Clone: RNL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR