Anti-E Tag 0 Polyclonal Antibody

Supplier: Abnova
PAB13742
10711-526EA 530.32 USD
10711-526
Anti-E Tag 0 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptides of E tag.

Recommended Dilutions: Western Blot: 0.2-1.0 ?g/ml; Immunofluorescence: 1.0-4.0 ?g/ml

Type: Primary
Antigen: E Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: 0
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR