Anti-DYKDDDDK tag Polyclonal Antibody

Supplier: Abnova
PAB13743
10713-358EA 530.32 USD
10713-358
Anti-DYKDDDDK tag Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptides of DYKDDDDK tag.

Recommended Dilutions: ELISA: Detection: 0.04-0.2 ?g/ml Capture: 5.0 ?g/ml; Western Blot: 0.2-1.0 ?g/ml

Type: Primary
Antigen: DYKDDDDK tag
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR