Anti-Glu-Glu (EYMPME) Polyclonal Antibody

Supplier: Abnova
PAB13745
10715-990EA 530.32 USD
10715-990
Anti-Glu-Glu (EYMPME) Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Glu-Glu.

Recommended Dilutions: ELISA: Detection: 1:1000-1:10000 Capture: 1:200-1:500; Western Blot: 1:500-1:2500

Type: Primary
Antigen: Glu-Glu (EYMPME)
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR