Anti-GST Polyclonal Antibody

Supplier: Abnova
PAB13747
10720-734EA 427.45 USD
10720-734
Anti-GST Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against GST.

Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:5000

Type: Primary
Antigen: GST
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR