Anti-Mycobacterium avium Mouse Monoclonal Antibody [clone: 103]

Supplier: Abnova
MAB4265
10681-158EA 2296.78 USD
10681-158
Anti-Mycobacterium avium Mouse Monoclonal Antibody [clone: 103]
Antibodies
Mouse monoclonal antibody raised against Mycobacterium avium.

Type: Primary
Antigen: Mycobacterium avium
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 103
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR