Anti-NEO1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22071
10726-934EA 428.55 USD
10726-934
Anti-NEO1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant NEO1.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: NEO1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR