Anti-PRL Mouse Monoclonal Antibody [clone: 090-18111]

Supplier: Abnova
MAB4301
10628-422EA 530.87 USD
10628-422
Anti-PRL Mouse Monoclonal Antibody [clone: 090-18111]
Antibodies
Mouse monoclonal antibody raised against native PRL.

Type: Primary
Antigen: PRL
Clonality: Monoclonal
Clone: 090-18111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR