Anti-PRL Mouse Monoclonal Antibody [clone: ME.121]

Supplier: Abnova
MAB4302
10638-298EA 4173.27 USD
10638-298
Anti-PRL Mouse Monoclonal Antibody [clone: ME.121]
Antibodies
Mouse monoclonal antibody raised against PRL.

Type: Primary
Antigen: PRL
Clonality: Monoclonal
Clone: ME.121
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR