Anti-PECAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-05]

Supplier: Abnova
MAB5045
10728-872EA 329 USD
10728-872
Anti-PECAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-05]
Antibodies
Mouse monoclonal antibody raised against native PECAM1.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: MEM-05
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR