Anti-BSG 0 Monoclonal Antibody (Biotin) [clone: MEM-M6/1]

Supplier: Abnova
MAB5048
10691-574EA 329 USD
10691-574
Anti-BSG 0 Monoclonal Antibody (Biotin) [clone: MEM-M6/1]
Antibodies
Mouse monoclonal antibody raised against recombinant BSG.

Recommended Dilutions: Flow Cytometry: 1:250

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: MEM-M6/1
Conjugation: Biotin
Epitope:
Host: 0
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR