Anti-HLA-G Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM-G/9]

Supplier: Abnova
MAB5058
10717-590EA 428 USD
10717-590
Anti-HLA-G Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM-G/9]
Antibodies
Mouse monoclonal antibody raised against recombinant HLA-G.

Recommended Dilutions: Flow Cytometry: 1:500

Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone: MEM-G/9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR