Anti-XAGE1D Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11978
10744-740EA 530.32 USD
10744-740
Anti-XAGE1D Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of XAGE1.

Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: XAGE1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR