Anti-C6orf162 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20377
10705-400EA 428.55 USD
10705-400
Anti-C6orf162 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C6orf162.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: C6orf162
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR