Anti-Influenza A virus Hemagglutinin H5 Mouse Monoclonal Antibody [clone: 1E7D8]

Supplier: Abnova
MAB2765
10719-350EA 530.32 USD
10719-350
Anti-Influenza A virus Hemagglutinin H5 Mouse Monoclonal Antibody [clone: 1E7D8]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of Influenza A Virus Haemagglutinin H5.

Recommended Dilutions: ELISA: 1 ?g/ml

Type: Primary
Antigen: Influenza A virus Hemagglutinin H5
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 1E7D8
Epitope:
Host: IgG
Isotype: Mouse
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR