Anti-Influenza A virus hemagglutinin HA1 Mouse Monoclonal Antibody [clone: 4E11E1]

Supplier: Abnova
MAB2768
10719-360EA 530.32 USD
10719-360
Anti-Influenza A virus hemagglutinin HA1 Mouse Monoclonal Antibody [clone: 4E11E1]
Antibodies
Mouse monoclonal antibody raised against recombinant Hemagglutinin.

Recommended Dilutions: Western Blot: 2.5-5 ?g/ml

Type: Primary
Antigen: Influenza A virus hemagglutinin HA1
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 4E11E1
Epitope:
Host: IgG
Isotype: Mouse
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR