Anti-GAPDH Mouse Monoclonal Antibody [clone: 6F7]

Supplier: Abnova
MAB2783
10682-430EA 427.45 USD
10682-430
Anti-GAPDH Mouse Monoclonal Antibody [clone: 6F7]
Antibodies
Mouse monoclonal antibody raised against GAPDH.

Type: Primary
Antigen: GAPDH
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 6F7
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR