Anti-ADIPOQ Mouse Monoclonal Antibody [clone: Adn2]

Supplier: Abnova
MAB2790
10673-112EA 428.55 USD
10673-112
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: Adn2]
Antibodies
Mouse monoclonal antibody raised against native ADIPOQ.

Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: Adn2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR