Anti-ADIPOQ Mouse Monoclonal Antibody [clone: Adn23]

Supplier: Abnova
MAB2791
10673-114EA 428.55 USD
10673-114
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: Adn23]
Antibodies
Mouse monoclonal antibody raised against native ADIPOQ.

Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: Adn23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR