Anti-ADIPOQ Mouse Monoclonal Antibody [clone: Adn27]

Supplier: Abnova
MAB2792
10673-116EA 428.55 USD
10673-116
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: Adn27]
Antibodies
Mouse monoclonal antibody raised against native ADIPOQ.

Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: Adn27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR