Anti-COL15A1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20976
10708-604EA 428.55 USD
10708-604
Anti-COL15A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant COL15A1.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: COL15A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR