Anti-EPHX1 Mouse Monoclonal Antibody [clone: AT2E5]

Supplier: Abnova
MAB3588
10712-250EA 329 USD
10712-250
Anti-EPHX1 Mouse Monoclonal Antibody [clone: AT2E5]
Antibodies
Mouse monoclonal antibody raised against partial recombinant EPHX1.

Type: Primary
Antigen: EPHX1
Clonality: Monoclonal
Clone: AT2E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR