Anti-BIK Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12962
10691-234EA 527.69 USD
10691-234
Anti-BIK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of BIK.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: BIK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR