Anti-EDA2R Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12963
10711-616EA 428 USD
10711-616
Anti-EDA2R Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody raised against recombinant EDA2R.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: EDA2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR