Anti-AIV NS1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12968
10690-580EA 428 USD
10690-580
Anti-AIV NS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Avian Influenza NS1.

Type: Primary
Antigen: AIV NS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR