Anti-N4BP2L2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23427
10726-502EA 428.55 USD
10726-502
Anti-N4BP2L2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant N4BP2L2.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: N4BP2L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR