Anti-TARBP1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12981
10740-516EA 428 USD
10740-516
Anti-TARBP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TARBP1.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: TARBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR