Anti-Transportin 3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23434
10742-316EA 428.55 USD
10742-316
Anti-Transportin 3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant TNPO3.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: Transportin 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR