Anti-PTPRC Mouse Monoclonal Antibody [clone: IVA103]

Supplier: Abnova
MAB3619
10706-612EA 329 USD
10706-612
Anti-PTPRC Mouse Monoclonal Antibody [clone: IVA103]
Antibodies
Mouse monoclonal antibody raised against native CD45R.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: IVA103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR