Anti-Kinesin (heavy chain) Mouse Monoclonal Antibody [clone: KN-02]

Supplier: Abnova
MAB3621
10722-470EA 329 USD
10722-470
Anti-Kinesin (heavy chain) Mouse Monoclonal Antibody [clone: KN-02]
Antibodies
Mouse monoclonal antibody raised against native Kinesin (heavy chain).

Type: Primary
Antigen: Kinesin (heavy chain)
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: KN-02
Epitope:
Host: IgM
Isotype: Mouse
Reactivity: Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR