Anti-FN1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13520
10714-964EA 653.85 USD
10714-964
Anti-FN1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against native Fn1.

Recommended Dilutions: ELISA: 1:200; Immunohistochemistry: 1:500-1:1000; Immunofluorescence: 1:80

Type: Primary
Antigen: Fn1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR