Anti-KLK3 Mouse Monoclonal Antibody [clone: 981]

Supplier: Abnova
MAB4311
10627-910EA 638.45 USD
10627-910
Anti-KLK3 Mouse Monoclonal Antibody [clone: 981]
Antibodies
Mouse monoclonal antibody raised against KLK3.

Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: 981
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR