Anti-Staphylococcus aureus Mouse Monoclonal Antibody [clone: 702]

Supplier: Abnova
MAB4339
10677-286EA 2827.69 USD
10677-286
Anti-Staphylococcus aureus Mouse Monoclonal Antibody [clone: 702]
Antibodies
Mouse monoclonal antibody raised against Staphylococcus aureus.

Type: Primary
Antigen: Staphylococcus aureus
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 702
Epitope:
Host: IgG3
Isotype: Mouse
Reactivity: Staphylococcus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR